accessibility menu, dialog, popup

UserName

CAS RN 15461-27-5

ZrOClCl

CAS RN 15461-27-5

IUPAC Name: oxozirconium hydrate dihydrochloride
Synonyms: oxozirconium hydrate dihydrochloride
Molecular Weight (g/mol): 198.15
Molecular Formula: Cl2H4O2Zr
InChi Key: VBTPZBJEHPOUOI-UHFFFAOYSA-N
SMILES: O.Cl.Cl.O=[Zr]
Molecular Weight (g/mol) 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)