accessibility menu, dialog, popup

UserName

H3CNH

CAS RN 1551-06-0

IUPAC Name: 2-ethyl-1H-pyrrole
Synonyms: ethazole
Molecular Weight (g/mol): 95.15
Molecular Formula: C6H9N
InChi Key: XRPDDDRNQJNHLQ-UHFFFAOYSA-N
SMILES: CCC1=CC=CN1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results