CAS RN 155601-30-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4,5-Diamino-1-(2-hydroxyethyl)pyrazole Sulfate, TRC
CAS: 155601-30-2 Molecular Formula: C5H10N4O . H2SO4 Molecular Weight (g/mol): 240.23 Synonym: 1-(2-Hydroxyethyl)-4,5-diaminopyrazole Sulfate, 4,5-Diamino-1-(2-hydroxyethyl)-1H-pyrazole Sulfate, Jarocol AHP, Rodol HDAP, 4,5-Diamino-1H-pyrazole-1-ethanol Sulfate (1:1) IUPAC Name: 2-(4,5-diamino-1H-pyrazol-1-yl)ethan-1-ol sulfate SMILES: Nc1c(N)n(CCO)nc1.OS(=O)(O)=O