accessibility menu, dialog, popup

UserName

NNClF

CAS RN 155937-28-3

IUPAC Name: 1-chloro-4-(4-fluorophenyl)phthalazine
Synonyms: 1-chloro-4-(4-fluorophenyl)phthalazine
Molecular Weight (g/mol): 258.68
Molecular Formula: C14H8ClFN2
InChi Key: LIFADULWFCVAQR-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=NN=C(Cl)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results