accessibility menu, dialog, popup

UserName

CAS RN 1562-85-2

IUPAC Name: [1-carboxy-7-(dimethylamino)-4-hydroxy-10H-phenoxazin-3-yl]oxidanylium chloride
Synonyms: [1-carboxy-7-(dimethylamino)-4-hydroxy-10H-phenoxazin-3-yl]oxidanylium chloride
Molecular Weight (g/mol): 336.73
Molecular Formula: C15H13ClN2O5
InChi Key: IQVJEUYKPJBCKY-UHFFFAOYSA-M
SMILES: [Cl-].CN(C)C1=CC=C2NC3=C(OC2=C1)C(O)=C([O+])C=C3C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results