accessibility menu, dialog, popup

UserName

CAS RN 15622-40-9

HoHoOSOOOOSOOOOSOOO

CAS RN 15622-40-9

IUPAC Name: diholmium(3+) trisulfate
Synonyms: diholmium(3+) trisulfate
Molecular Weight (g/mol): 618.03
Molecular Formula: Ho2O12S3
InChi Key: MKPJADFELTTXAV-UHFFFAOYSA-H
SMILES: [Ho+3].[Ho+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results