accessibility menu, dialog, popup

UserName

CAS RN 15632-39-0

YOOOOOOCCCCH3H3CH3CCH3CH3CH3CH3H3CCH3H3CH3CCH3CH3CH3H3CH3CCH3H3CHHH

CAS RN 15632-39-0

IUPAC Name: tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) yttrium
Synonyms: tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) yttrium
Molecular Weight (g/mol): 641.74
Molecular Formula: C33H60O6Y
InChi Key: IAYLRLKRHZVTOP-UHFFFAOYSA-N
SMILES: [Y].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results