accessibility menu, dialog, popup

UserName

CH3OOOFFFF

CAS RN 156783-98-1

IUPAC Name: methyl 2,2,3,3-tetrafluoropropyl carbonate
Synonyms: methyl 2,2,3,3-tetrafluoropropyl carbonate
Molecular Weight (g/mol): 190.09
Molecular Formula: C5H6F4O3
InChi Key: LDTVHHNIXCBROE-UHFFFAOYSA-N
SMILES: COC(=O)OCC(F)(F)C(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results