accessibility menu, dialog, popup

UserName

CAS RN 158849-23-1

CH3(R)H3NOOCH3CH3H3C

CAS RN 158849-23-1

IUPAC Name: (2R)-4-(tert-butoxy)-4-oxobutan-2-aminium
Synonyms: (2R)-4-(tert-butoxy)-4-oxobutan-2-aminium
Molecular Weight (g/mol): 160.24
Molecular Formula: C8H18NO2
InChi Key: BFFNZGWJTHWUMY-ZCFIWIBFSA-O
SMILES: C[C@@H]([NH3+])CC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)