accessibility menu, dialog, popup

UserName

CAS RN 15893-42-2

IUPAC Name: 3-(4-methoxyphenyl)propanoyl chloride
Synonyms: 3-(4-methoxyphenyl)propanoyl chloride
Molecular Weight (g/mol): 198.65
Molecular Formula: C10H11ClO2
InChi Key: FQVJPHCAWYRYCK-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCC(Cl)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results