CAS RN 158985-37-6
IUPAC Name:
tert-butyl 4-[4-(hydroxymethyl)phenyl]piperazine-1-carboxylate
Synonyms:
tert-butyl 4-[4-(hydroxymethyl)phenyl]piperazine-1-carboxylate
Molecular Weight (g/mol):
292.38
Molecular Formula:
C16H24N2O3
InChi Key:
PXKYXTXYWJKTSQ-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(CO)C=C1
1
–
1
of
1
results
1
tert-Butyl 4-[4-(hydroxymethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate, ≥97%, Thermo Scientific™
CAS: 158985-37-6 Molecular Formula: C16H24N2O3 Molecular Weight (g/mol): 292.379 MDL Number: MFCD06411543 InChI Key: PXKYXTXYWJKTSQ-UHFFFAOYSA-N Synonym: tert-butyl 4-4-hydroxymethyl phenyl piperazine-1-carboxylate,4-4-n-boc-piperazinyl benzyl alcohol,tert-butyl 4-4-hydroxymethyl phenyl tetrahydro-1 2h-pyrazinecarboxylate,4-4-n-boc-piperazinyl benzylalcohol,4-4-tert-butoxycarbonyl piperazine benzyl alcohol,pubchem11318,buttpark 98\06-99,4-1-boc-piperazin-4-yl phenylmethanol,4-4-tert-butoxycarbonyl piperazin-1-yl benzyl alcohol PubChem CID: 2795510 IUPAC Name: tert-butyl 4-[4-(hydroxymethyl)phenyl]piperazine-1-carboxylate SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)CO