CAS RN 159001-25-9
CAS RN 159001-25-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dihydrocephalomannine, TRC
CAS: 159001-25-9 Molecular Formula: C45 H55 N O14 Molecular Weight (g/mol): 833.92 Synonym: 2'',3''-Dihydrocephalomannine,N-Debenzoyl-N-α-methylbutyrylpaclitaxel SMILES: CCC(C)C(=O)N[C@H]([C@@H](O)C(=O)O[C@H]1C[C@@]2(O)[C@@H](OC(=O)c3ccccc3)[C@@H]4[C@@]5(CO[C@@H]5C[C@H](O)[C@@]4(C)C(=O)[C@H](OC(=O)C)C(=C1C)C2(C)C)OC(=O)C)c6ccccc6