accessibility menu, dialog, popup

UserName

NOOH

CAS RN 159691-25-5

IUPAC Name: 4-[(pyrrolidin-1-yl)methyl]benzoic acid
Synonyms: 4-(pyrrolidin-1-ylmethyl)benzoic acid
Molecular Weight (g/mol): 205.26
Molecular Formula: C12H15NO2
InChi Key: DOEIPZUTRVMWAB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(CN2CCCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results