accessibility menu, dialog, popup

UserName

ONH2N

CAS RN 159724-40-0

IUPAC Name: 3-(morpholin-4-yl)aniline
Synonyms: 3-(morpholin-4-yl)aniline
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: ZJWLMZURLIHVHE-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)N1CCOCC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results