accessibility menu, dialog, popup

UserName

CAS RN 15985-39-4

H3CSHNO(S)NH3OO

CAS RN 15985-39-4

IUPAC Name: (2S)-2-azaniumyl-4-[imino(methyl)oxo-λ⁶-sulfanyl]butanoate
Synonyms: (2S)-2-ammonio-4-[imino(methyl)oxo-λ⁶-sulfanyl]butanoate
Molecular Weight (g/mol): 180.22
Molecular Formula: C5H12N2O3S
InChi Key: SXTAYKAGBXMACB-DPVSGNNYSA-N
SMILES: CS(=N)(=O)CC[C@H]([NH3+])C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)