CAS RN 1603-55-0
CAS RN 1603-55-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Cyclohexene-d10, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
Cyclohexene-d10, TRC
CAS: 1603-55-0 Molecular Formula: C6 D10 Molecular Weight (g/mol): 92.21 Synonym: Cyclohexene-1,2,3,3,4,4,5,5,6,6-d10 IUPAC Name: 1,2,3,3,4,4,5,5,6,6-decadeuteriocyclohexene SMILES: [2H]C1=C([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]