accessibility menu, dialog, popup

UserName

CH3SNOHNFFF

CAS RN 16097-62-4

IUPAC Name: 2-(methylsulfanyl)-6-(trifluoromethyl)-1,4-dihydropyrimidin-4-one
Synonyms: 2-(methylsulfanyl)-6-(trifluoromethyl)-1H-pyrimidin-4-one
Molecular Weight (g/mol): 210.17
Molecular Formula: C6H5F3N2OS
InChi Key: BRLPDRZKOMZVOY-UHFFFAOYSA-N
SMILES: CSC1=NC(=O)C=C(N1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results