accessibility menu, dialog, popup

UserName

CH3CH3CH3H3CCH3NCO

CAS RN 1611-57-0

IUPAC Name: 2-isocyanato-2,4,4-trimethylpentane
Synonyms: 2-isocyanato-2,4,4-trimethylpentane
Molecular Weight (g/mol): 155.24
Molecular Formula: C9H17NO
InChi Key: XOHBENIMDRFUIH-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results