accessibility menu, dialog, popup

UserName

CH3NHNNH2

CAS RN 16153-81-4

IUPAC Name: 4-(4-aminophenyl)-1-methylpiperazin-1-ium
Synonyms: 4-(4-aminophenyl)-1-methylpiperazin-1-ium
Molecular Weight (g/mol): 192.29
Molecular Formula: C11H18N3
InChi Key: MOZNZNKHRXRLLF-UHFFFAOYSA-O
SMILES: C[NH+]1CCN(CC1)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)