CAS RN 161618-30-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Chloro-3-iodo Hydroxychloroquine, TRC
CAS: 161618-30-0 Molecular Formula: C18H25ClIN3O Molecular Weight (g/mol): 461.77 Synonym: 2-((4-((7-Chloro-3-iodoquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol IUPAC Name: 2-[4-[(7-chloro-3-iodoquinolin-4-yl)amino]pentyl-ethylamino]ethanol SMILES: CCN(CCO)CCCC(NC1=C2C=CC(Cl)=CC2=NC=C1I)C