accessibility menu, dialog, popup

UserName

CAS RN 162046-61-9

FFFOClO

CAS RN 162046-61-9

IUPAC Name: 2-(trifluoromethoxy)benzoyl chloride
Synonyms: 2-(trifluoromethoxy)benzoyl chloride
Molecular Weight (g/mol): 224.56
Molecular Formula: C8H4ClF3O2
InChi Key: KSHPWYXAJJCUMJ-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC=C1C(Cl)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)