accessibility menu, dialog, popup

UserName

CAS RN 16205-90-6

CH3OOH3C

CAS RN 16205-90-6

IUPAC Name: ethyl hex-2-ynoate
Synonyms: ethyl hex-2-ynoate
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: ZFMNCZDBAHELNX-UHFFFAOYSA-N
SMILES: CCCC#CC(=O)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results