accessibility menu, dialog, popup

UserName

CAS RN 162101-31-7

IUPAC Name: (2-fluoro-4-methoxyphenyl)boronic acid
Synonyms: 2-fluoro-4-methoxyphenylboronic acid
Molecular Weight (g/mol): 169.95
Molecular Formula: C7H8BFO3
InChi Key: ULUIXJDBPYBAHS-UHFFFAOYSA-N
SMILES: COC1=CC=C(B(O)O)C(F)=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results