accessibility menu, dialog, popup

UserName

H3COOHN

CAS RN 162402-33-7

IUPAC Name: 4-(4-methoxyphenoxy)piperidine
Synonyms: 4-(4-methoxyphenoxy)piperidine
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: HRMYEAQYLDCUGG-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC2CCNCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results