accessibility menu, dialog, popup

UserName

CH3NH3CBBNH3CCH3NH3CCH3NCH3CH3

CAS RN 1630-79-1

IUPAC Name: dimethyl[1,2,2-tris(dimethylamino)diboran-1-yl]amine
Synonyms: dimethyl[1,2,2-tris(dimethylamino)diboran-1-yl]amine
Molecular Weight (g/mol): 197.93
Molecular Formula: C8H24B2N4
InChi Key: KMCDRSZVZMXKRL-UHFFFAOYSA-N
SMILES: CN(C)B(B(N(C)C)N(C)C)N(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)