accessibility menu, dialog, popup

UserName

H3CNN

CAS RN 1632-83-3

IUPAC Name: 1-methyl-1H-1,3-benzodiazole
Synonyms: 1-methyl-1,3-benzodiazole
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: FGYADSCZTQOAFK-UHFFFAOYSA-N
SMILES: CN1C=NC2=CC=CC=C12
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)