CAS RN 1632051-40-1
CAS RN 1632051-40-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Zuranolone, MedChemExpress
MedChemExpress Zuranolone is an orally active and potent neuroactive steroid positive allosteric modulator of GABAA receptor, with EC50s of 296 and 163 nM for α1β2γ2 and α4β3δ GABAA receptors, respectively.