accessibility menu, dialog, popup

UserName

HNFFHCl

CAS RN 163457-23-6

IUPAC Name: 3,3-difluoropyrrolidine hydrochloride
Synonyms: 3,3-difluoropyrrolidine hydrochloride
Molecular Weight (g/mol): 143.56
Molecular Formula: C4H8ClF2N
InChi Key: YYVPZQADFREIFR-UHFFFAOYSA-N
SMILES: Cl.FC1(F)CCNC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results