CAS RN 163759-49-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2'-O-(2-Methoxyethyl)-5-methyl-uridine, TRC
CAS: 163759-49-7 Molecular Formula: C13H20N2O7 Molecular Weight (g/mol): 316.31 IUPAC Name: 1-[(2R,3R,4R,5R)-4-hydroxy-5-(hydroxymethyl)-3-(2-methoxyethoxy)oxolan-2-yl]-5-methylpyrimidine-2,4-dione SMILES: COCCO[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N2C=C(C)C(=O)NC2=O