CAS RN 1638785-17-7
CAS RN 1638785-17-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(2-Chloroethyl)-N-methoxy-N,Alpha,Alpha-trimethyl-benzeneacetamide, TRC
CAS: 1638785-17-7 Molecular Formula: C14 H20 Cl N O2 Molecular Weight (g/mol): 269.7671 Synonym: 4-(2-Chloroethyl)-N-methoxy-N,α,α-trimethylbenzeneacetamide,2-(4-(3-Chloropropanoyl)phenyl)-N-methoxy-N,2-dimethylpropanamide IUPAC Name: 2-[4-(2-chloroethyl)phenyl]-N-methoxy-N,2-dimethylpropanamide SMILES: CON(C)C(=O)C(C)(C)c1ccc(CCCl)cc1