accessibility menu, dialog, popup

UserName

CAS RN 16454-60-7

H2OH2OH2OH2OH2OH2ONdONOOONOOONOO

CAS RN 16454-60-7

IUPAC Name: neodymium(3+) hexahydrate trinitrate
Synonyms: neodymium(3+) hexahydrate trinitrate
Molecular Weight (g/mol): 438.34
Molecular Formula: H12N3NdO15
InChi Key: VQVDTKCSDUNYBO-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Nd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results