accessibility menu, dialog, popup

UserName

NH2NHNBr

CAS RN 16461-94-2

IUPAC Name: 4-bromo-1H-pyrazol-5-amine
Synonyms: 4-bromo-2H-pyrazol-3-amine
Molecular Weight (g/mol): 161.99
Molecular Formula: C3H4BrN3
InChi Key: OELYMZVJDKSMOJ-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=NN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results