accessibility menu, dialog, popup

UserName

CH3SI

CAS RN 16494-36-3

IUPAC Name: 2-iodo-5-methylthiophene
Synonyms: 2-iodo-5-methylthiophene
Molecular Weight (g/mol): 224.06
Molecular Formula: C5H5IS
InChi Key: NAZNQEXKAPLVKC-UHFFFAOYSA-N
SMILES: CC1=CC=C(I)S1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results