accessibility menu, dialog, popup

UserName

CAS RN 165324-09-4

IUPAC Name: calcium bis(trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane)
Synonyms: calcium bis(bistriflylimide anion)
Molecular Weight (g/mol): 600.35
Molecular Formula: C4CaF12N2O8S4
InChi Key: WFABOCFDABTAPE-UHFFFAOYSA-N
SMILES: [Ca++].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)