accessibility menu, dialog, popup

UserName

CAS RN 16533-72-5

OO(R)O(S)OO

CAS RN 16533-72-5

IUPAC Name: (2R,3S)-oxirane-2,3-dicarboxylate
Synonyms: (2R,3S)-oxirane-2,3-dicarboxylate
Molecular Weight (g/mol): 130.06
Molecular Formula: C4H2O5
InChi Key: DCEMCPAKSGRHCN-XIXRPRMCSA-L
SMILES: [O-]C(=O)[C@H]1O[C@H]1C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)