accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 1654-86-0

IUPAC Name: decyl decanoate
Synonyms: decanoic acid, decyl ester
Molecular Weight (g/mol): 312.54
Molecular Formula: C20H40O2
InChi Key: XAKXZZPEUKNHMA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCOC(=O)CCCCCCCCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results