accessibility menu, dialog, popup

UserName

CH3NH3CH3CNCH3

CAS RN 1660-93-1

IUPAC Name: 3,4,7,8-tetramethyl-1,10-phenanthroline
Synonyms: 3,4,7,8-tetramethyl-1,10-phenanthroline
Molecular Weight (g/mol): 236.32
Molecular Formula: C16H16N2
InChi Key: NPAXPTHCUCUHPT-UHFFFAOYSA-N
SMILES: CC1=CN=C2C(C=CC3=C(C)C(C)=CN=C23)=C1C
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)