accessibility menu, dialog, popup

UserName

CH3CH3H3COOH2C

CAS RN 1663-39-4

IUPAC Name: tert-butyl prop-2-enoate
Synonyms: tert-butyl prop-2-enoate
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: ISXSCDLOGDJUNJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)C=C
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)