accessibility menu, dialog, popup

UserName

CAS RN 1669-44-9

CH3CH3O(E)

CAS RN 1669-44-9

IUPAC Name: (3E)-oct-3-en-2-one
Synonyms: (3E)-oct-3-en-2-one
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: ZCFOBLITZWHNNC-VOTSOKGWSA-N
SMILES: CCCC\C=C\C(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results