accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHOH

CAS RN 167081-25-6

IUPAC Name: tert-butyl N-(4-hydroxycyclohexyl)carbamate
Synonyms: tert-butyl N-(4-hydroxycyclohexyl)carbamate
Molecular Weight (g/mol): 215.29
Molecular Formula: C11H21NO3
InChi Key: DQARDWKWPIRJEH-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(O)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results