CAS RN 167223-43-0
CAS RN 167223-43-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
D-tert-Leucine Methyl Ester Hydrochloride, TRC
CAS: 167223-43-0 Molecular Formula: C7H15NO2 . HCl Molecular Weight (g/mol): 181.66 IUPAC Name: methyl (2R)-2-amino-3,3-dimethylbutanoate;hydrochloride SMILES: Cl.COC(=O)[C@H](N)C(C)(C)C