CAS RN 16844-71-6
CAS RN 16844-71-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Epifriedelanol, TRC
CAS: 16844-71-6 Molecular Formula: C30H52O Molecular Weight (g/mol): 428.73 Synonym: (3β)-D:A-Friedooleanan-3-ol,D:A-Friedooleanan-3β-ol,3-Epifriedelinol,3-epi-Friedanol,3β-Hydroxyfriedelane,Epifriedelanol,Epifriedelinol,Friedelan-3β-ol,Friedelin-3β-ol,Longan Triterpane A,epi-Friedelanol,β-Friedelinol IUPAC Name: (3S,4R,4aS,6bR,8aR,12aR,12bS,14aS,14bS)-4,4a,6b,8a,11,11,12b,14a-octamethyldocosahydropicen-3-ol SMILES: [H][C@@]12[C@@](CCC(C)(C)C2)(C)CC[C@]3(C)C4CC[C@]5(C)[C@@H](C)[C@@H](O)CC[C@@]5([H])[C@]4(C)CC[C@]31C