CAS RN 16853-74-0
IUPAC Name:
zirconium(4+) tungsten pentaoxidandiide
Synonyms:
zirconium tungstate
Molecular Weight (g/mol):
355.06
Molecular Formula:
O5WZr
InChi Key:
GGUDROIMBGREID-UHFFFAOYSA-N
SMILES:
[O--].[O--].[O--].[O--].[O--].[Zr+4].[W]
1
–
1
of
1
results
1
Tungsten zirconium oxide, 99.7% (metals basis excluding Hf), Hf <0.38%, Thermo Scientific Chemicals
CAS: 16853-74-0 MDL Number: MFCD00168096 Synonym: Zirconium tungstate; Zirconium tungsten oxide