accessibility menu, dialog, popup

UserName

CAS RN 169-14-2

ClHNOONH

CAS RN 169-14-2

IUPAC Name: 6-chloro-1,2,3,4-tetrahydroquinoxaline-2,3-dione
Synonyms: 6-chloro-1,4-dihydroquinoxaline-2,3-dione
Molecular Weight (g/mol): 196.59
Molecular Formula: C8H5ClN2O2
InChi Key: RNOLFZACEWWIHP-UHFFFAOYSA-N
SMILES: ClC1=CC=C2NC(=O)C(=O)NC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results