accessibility menu, dialog, popup

UserName

CH3H3CCH3ONHNOH

CAS RN 169205-93-0

IUPAC Name: N-(3-hydroxypyridin-4-yl)-2,2-dimethylpropanamide
Synonyms: N-(3-hydroxypyridin-4-yl)-2,2-dimethylpropanamide
Molecular Weight (g/mol): 194.23
Molecular Formula: C10H14N2O2
InChi Key: HZNFSZMIMUTCGG-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)NC1=C(O)C=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results