CAS RN 16962-47-3
CAS RN 16962-47-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
230.76
Molecular Formula:
F6GeH16N2
InChi Key:
LHKQHMLGVWLMTF-UHFFFAOYSA-N
SMILES:
[NH4+].[NH4+].F.F.F.F.[F-].[F-].[GeH4]
1
–
1
of
1
results
1
Ammonium hexafluorogermanate, 99.99% min (metals basis)
CAS: 16962-47-3 Molecular Formula: H8F6GeN2 MDL Number: MFCD00010880