accessibility menu, dialog, popup

UserName

CAS RN 169677-20-7

H3CH3CCH3SOOCl

CAS RN 169677-20-7

IUPAC Name: 4-(2-methylbutan-2-yl)benzene-1-sulfonyl chloride
Synonyms: 4-(2-methylbutan-2-yl)benzenesulfonyl chloride
Molecular Weight (g/mol): 246.75
Molecular Formula: C11H15ClO2S
InChi Key: WLBQFZZPRWQTGR-UHFFFAOYSA-N
SMILES: CCC(C)(C)C1=CC=C(C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results