accessibility menu, dialog, popup

UserName

O

CAS RN 1708-29-8

IUPAC Name: 2,5-dihydrofuran
Synonyms: 2,5-dihydrofuran
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: ARGCQEVBJHPOGB-UHFFFAOYSA-N
SMILES: C1OCC=C1
Quantity 
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)