accessibility menu, dialog, popup

UserName

BOOCH3NH2

CAS RN 17093-22-0

IUPAC Name: diphenylboranyl 2-aminopentanoate
Synonyms: diphenylboranyl 2-aminopentanoate
Molecular Weight (g/mol): 281.16
Molecular Formula: C17H20BNO2
InChi Key: XAVUFSLMIUPEFM-UHFFFAOYNA-N
SMILES: CCCC(N)C(=O)OB(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results