accessibility menu, dialog, popup

UserName

H3COOOCH3CH3H3C

CAS RN 17094-34-7

IUPAC Name: ethyl 4,4-dimethyl-3-oxopentanoate
Synonyms: ethyl 4,4-dimethyl-3-oxopentanoate
Molecular Weight (g/mol): 172.22
Molecular Formula: C9H16O3
InChi Key: VUYNTIDSHCJIKF-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results